missing translation for 'onlineSavingsMsg'
Learn More

3-n-Butoxypropionitrile, 97%, Thermo Scientific™

Product Code. 11397716
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6959-71-3
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.19
MDL Number MFCD00045643
InChI Key AWVNJBFNHGQUQU-UHFFFAOYSA-N
Synonym 3-butoxypropionitrile, propanenitrile, 3-butoxy, beta-butoxy propionitrile, propionitrile, 3-butoxy, 3-butoxypropannitril, 3-butoxypropiononitrile, 3-n-butoxypropionitrile, 3-butoxypropannitril czech, .beta.-butoxy propionitrile, 3-butoxypropanitrile
PubChem CID 23395
IUPAC Name 3-butoxypropanenitrile
SMILES CCCCOCCC#N

Specifications

Density 0.92
Boiling Point 105°C to 106°C (15 mmHg)
Flash Point 89°C (192°F)
Refractive Index 1.4175
Quantity 5 g
Beilstein 1747904
Formula Weight 127.19
Percent Purity 97%
Chemical Name or Material 3-n-Butoxypropionitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'