missing translation for 'onlineSavingsMsg'
Learn More
3-Nitro-D-phenylalanine, 95%, Thermo Scientific Chemicals Product Code.: 15480199

3-Nitro-D-phenylalanine, 95%, Thermo Scientific Chemicals

Product Code. 15480199
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15480199

Brand: Thermo Scientific Alfa Aesar H63483.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169530-97-6
Molecular Formula C9H10N2O4
Molecular Weight (g/mol) 210.19
MDL Number MFCD04117829
InChI Key YTHDRUZHNYKZGF-UHFFFAOYNA-N
Synonym r-2-amino-3-3-nitrophenyl propanoic acid, 3-nitro-d-phenylalanine, d-3-nitrophenylalanine, 2r-2-amino-3-3-nitrophenyl propanoic acid, d-3-no2-phe-oh, d-3-nitrophe, d-3-nitro phenylalanine, r-2-amino-3-3-nitrophenyl propanoicacid
PubChem CID 7000156
IUPAC Name 2-amino-3-(3-nitrophenyl)propanoic acid
SMILES NC(CC1=CC(=CC=C1)[N+]([O-])=O)C(O)=O

Specifications

Color White to Yellow
Quantity 250 mg
Formula Weight 210.19
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Nitro-D-phenylalanine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81301681 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH63483.03,H63483.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Nitro-D-phenylalanine, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.