missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
500 g
1000 g
Description
3-Nitrobenzaldehyde is used in the synthesis of other organic compounds, ranging from pharmaceuticals to plastic additives and intermediate for the processing of perfume and flavoring compounds and in the preparation of certain aniline dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Nitrobenzaldehyde is used in the synthesis of other organic compounds, ranging from pharmaceuticals to plastic additives and intermediate for the processing of perfume and flavoring compounds and in the preparation of certain aniline dyes.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
3-Nitrobenzaldehyde is used in the synthesis of other organic compounds, ranging from pharmaceuticals to plastic additives and intermediate for the processing of perfume and flavoring compounds and in the preparation of certain aniline dyes.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | 57°C to 59°C |
Boiling Point | 164°C (23 mmHg) |
Quantity | 1000 g |
Beilstein | 386795 |
Sensitivity | Air Sensitive |
Merck Index | 14,6587 |
Solubility Information | Slightly soluble in water. |
SMILES | C1=CC(=CC(=C1)[N+](=O)[O-])C=O |
Molecular Weight (g/mol) | 151.121 |
Formula Weight | 151.12 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'