missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 11490953
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13331-27-6
Molecular Formula C6H6BNO4
Molecular Weight (g/mol) 166.93
MDL Number MFCD00007193
InChI Key ZNRGSYUVFVNSAW-UHFFFAOYSA-N
Synonym 3-nitrophenyl boronic acid, 3-nitrobenzeneboronic acid, m-nitrophenylboronic acid, boronic acid, 3-nitrophenyl, m-nitrobenzeneboronic acid, benzeneboronic acid, m-nitro, nitrophenylboronic acid, m-nitophenyl boronic acid, 3-nitrophenyl boranediol
PubChem CID 1677
IUPAC Name (3-nitrophenyl)boronic acid
SMILES OB(O)C1=CC=CC(=C1)[N+]([O-])=O

Specifications

Melting Point ∼285°C (decomposition)
Quantity 25 g
Beilstein 2938638
Formula Weight 166.93
Percent Purity 98%
Chemical Name or Material 3-Nitrobenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'