missing translation for 'onlineSavingsMsg'
Learn More
3-Nitrobenzo[b]furan-5-ol, 97%, Thermo Scientific™ Product Code.: 10743823

3-Nitrobenzo[b]furan-5-ol, 97%, Thermo Scientific™

Product Code. 10743823
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 10743823

Brand: Thermo Scientific Maybridge SEW03534DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 126318-27-2
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C8H5NO4
MDL Number MFCD00114881
Quantity 1 g
Synonym 3-nitrobenzo b furan-5-ol, 5-benzofuranol,3-nitro, acmc-20e4do, 3-nitro-5-benzofuranol, 3-nitrobenzofuran-5-ol, 3-nitro-5-hydroxybenzofuran, 5-hydroxy-3-nitrobenzo b furan, 5-hydroxy-3-nitro-1-benzofuran
InChI Key ZQPBTBWGCWHIGU-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1O)C(=CO2)[N+](=O)[O-]
IUPAC Name 3-nitro-1-benzofuran-5-ol
Molecular Weight (g/mol) 179.131
PubChem CID 2797494
Formula Weight 179.13
Percent Purity 97%
Chemical Name or Material 3-Nitrobenzo[b]furan-5-ol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Nitrobenzo[b]furan-5-ol, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.