missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzoic Acid, 98%, Thermo Scientific Chemicals

Product Code. 10014623
Click to view available options
Quantity:
100 g
500 g
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-92-6
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007251
InChI Key AFPHTEQTJZKQAQ-UHFFFAOYSA-N
Synonym 3-nitrobenzoic acid, m-nitrobenzoic acid, benzoic acid, 3-nitro, benzoic acid, m-nitro, meta-nitrobenzoic acid, metanitrobenzoic acid, m-nitrobenzenecarboxylic acid, 3-nitro-benzoic acid, ccris 2335, nitrobenzoic 3-acid
PubChem CID 8497
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 139.0°C to 143.0°C
Color Green to Yellow
Flash Point 190°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula O2NC6H4CO2H
Quantity 100 g
Beilstein 09,376
Fieser 13,231
Merck Index 14,6589
Solubility Information Solubility in water: 3g/L (25°C). Other solubilities: 1g/3mL alcohol - 1g/2mL methanol, 1g/4mL ether - 1g/2.5mL acetone, 1g/18mL chloroform, slightly soluble in benzene and carbon disulfide
Formula Weight 167.12
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 3-Nitrobenzoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'