missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrotoluene, 99%, Thermo Scientific Chemicals

Product Code. 10563291
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-08-1
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007265
InChI Key QZYHIOPPLUPUJF-UHFFFAOYSA-N
Synonym 3-nitrotoluene, m-nitrotoluene, 3-nitrotoluol, benzene, 1-methyl-3-nitro, m-methylnitrobenzene, toluene, m-nitro, meta-nitrotoluol, 3-methylnitrobenzene, m-nitrotoluol, nitrotoluene, m
PubChem CID 7422
ChEBI CHEBI:39931
IUPAC Name 1-methyl-3-nitrobenzene
SMILES CC1=CC=CC(=C1)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 15°C to 16°C
Color Brown-Yellow to Yellow
Density 1.1500g/mL
Boiling Point 230°C to 231°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4NO2
Refractive Index 1.5450 to 1.547
Quantity 5 mL
Beilstein 05, 321
Merck Index 15, 6737
Specific Gravity 1.15
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and benzene,miscible with ether
Formula Weight 137.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Nitrotoluene, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Nitrotoluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'