missing translation for 'onlineSavingsMsg'
Learn More
3-Octyn-1-ol, 96%, Thermo Scientific Chemicals Product Code.: 11387435

3-Octyn-1-ol, 96%, Thermo Scientific Chemicals

Product Code. 11387435
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11387435

Brand: Thermo Scientific Alfa Aesar L05414.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Octyn-1-ol, is used for a study of the reduction of acetylenic compounds, including this alcohol, to (E)-alkenes by alkali metals in liquid ammonia. Some other applications includes as corrosion inhibitor formulation, metal complex solution, solvent stabilizer, biodegradable chemical intermediate to make pharmaceuticals, chemical compound synthesis and reagents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Octyn-1-ol, is used for a study of the reduction of acetylenic compounds, including this alcohol, to (E)-alkenes by alkali metals in liquid ammonia. Some other applications includes as corrosion inhibitor formulation, metal complex solution, solvent stabilizer, biodegradable chemical intermediate to make pharmaceuticals, chemical compound synthesis and reagents

Solubility
It is soluble in water.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14916-80-4
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00039552
InChI Key LRZGRGVRZSDRTK-UHFFFAOYSA-N
Synonym 3-octyn-1-ol, 3-octyne-1-ol, pubchem13072, acmc-1c9k2, 3-octyn-1-ol 5g
PubChem CID 84694
IUPAC Name oct-3-yn-1-ol
SMILES CCCCC#CCCO

Specifications

Density 0.883
Boiling Point 86°C to 88°C (9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.454
Quantity 25 g
Beilstein 1739950
Solubility Information It is soluble in water.
Formula Weight 126.2
Percent Purity 96%
Chemical Name or Material 3-Octyn-1-ol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10233255 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL05414.14,L05414.06
Lot Number10110769 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL05414.14,L05414.06
Lot NumberP24G004 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL05414.14,L05414.06
Lot NumberN15D035 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL05414.14,L05414.06
Lot Number10158225 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL05414.14,L05414.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Octyn-1-ol, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.