Learn More
3-Octyn-1-ol, 96%, Thermo Scientific Chemicals
57.18 € - 190.95 €
Chemical Identifiers
CAS | 14916-80-4 |
---|---|
Molecular Formula | C8H14O |
Molecular Weight (g/mol) | 126.199 |
MDL Number | MFCD00039552 |
InChI Key | LRZGRGVRZSDRTK-UHFFFAOYSA-N |
Synonym | 3-octyn-1-ol, 3-octyne-1-ol, pubchem13072, acmc-1c9k2, 3-octyn-1-ol 5g |
PubChem CID | 84694 |
IUPAC Name | oct-3-yn-1-ol |
SMILES | CCCCC#CCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11377435
|
Thermo Scientific Alfa Aesar
L05414.06 |
5 g |
57.18 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387435
|
Thermo Scientific Alfa Aesar
L05414.14 |
25 g |
190.95 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Octyn-1-ol, is used for a study of the reduction of acetylenic compounds, including this alcohol, to (E)-alkenes by alkali metals in liquid ammonia. Some other applications includes as corrosion inhibitor formulation, metal complex solution, solvent stabilizer, biodegradable chemical intermediate to make pharmaceuticals, chemical compound synthesis and reagents
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Octyn-1-ol, is used for a study of the reduction of acetylenic compounds, including this alcohol, to (E)-alkenes by alkali metals in liquid ammonia. Some other applications includes as corrosion inhibitor formulation, metal complex solution, solvent stabilizer, biodegradable chemical intermediate to make pharmaceuticals, chemical compound synthesis and reagents
Solubility
It is soluble in water.
Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Chemical Identifiers
14916-80-4 | |
126.199 | |
LRZGRGVRZSDRTK-UHFFFAOYSA-N | |
84694 | |
CCCCC#CCCO |
C8H14O | |
MFCD00039552 | |
3-octyn-1-ol, 3-octyne-1-ol, pubchem13072, acmc-1c9k2, 3-octyn-1-ol 5g | |
oct-3-yn-1-ol |
Specifications
14916-80-4 | |
86°C to 88°C (9 mmHg) | |
C8H14O | |
MFCD00039552 | |
3-octyn-1-ol, 3-octyne-1-ol, pubchem13072, acmc-1c9k2, 3-octyn-1-ol 5g | |
LRZGRGVRZSDRTK-UHFFFAOYSA-N | |
oct-3-yn-1-ol | |
84694 | |
96% |
0.883 | |
>110°C (230°F) | |
1.454 | |
1739950 | |
It is soluble in water. | |
CCCCC#CCCO | |
126.199 | |
126.2 | |
3-Octyn-1-ol |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 238-986-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 3-Octyn-1-ol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 3-Octyn-1-ol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.