missing translation for 'onlineSavingsMsg'
Learn More
3-Oxetanol, 95%, Thermo Scientific Chemicals Product Code.: 15494318

3-Oxetanol, 95%, Thermo Scientific Chemicals

Product Code. 15494318
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15494318

Brand: Thermo Scientific Alfa Aesar H57928.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7748-36-9
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD09056790
InChI Key QMLWSAXEQSBAAQ-UHFFFAOYSA-N
Synonym 3-hydroxyoxetane, 3-oxetanol, 1,3-epoxy-2-propanol, oxetane-3-ol, 3-hydroxy-oxetane, 3-hydroxy oxetane, pubchem17367, ksc380k3t, 1,3-epoxypropan-2-olShow MoreShow Less
PubChem CID 9942117
IUPAC Name oxetan-3-ol
SMILES OC1COC1

Specifications

Density 1.167
Boiling Point 153°C
Flash Point 87°C (189°F)
Refractive Index 1.438
Quantity 5 g
Formula Weight 74.08
Percent Purity 95%
Chemical Name or Material 3-Oxetanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ23K018 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog NumberH57928.06,H57928.03
Lot NumberZ16H028 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH57928.06,H57928.03
Lot NumberZ30F046 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57928.06,H57928.03
Lot NumberP06A050 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57928.06,H57928.03
Lot NumberD06Y019 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57928.06,H57928.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Oxetanol, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.