Learn More
3-Oxo-1-cyclopentanecarboxylic acid, 97%, Thermo Scientific Chemicals
148.00 € - 515.00 €
Chemical Identifiers
CAS | 98-78-2 |
---|---|
Molecular Formula | C6H8O3 |
Molecular Weight (g/mol) | 128.13 |
MDL Number | MFCD01320173 |
InChI Key | RDSNBKRWKBMPOP-UHFFFAOYNA-N |
Synonym | 3-oxocyclopentanecarboxylic acid, 3-oxo-cyclopentane-carboxylic acid, 3-oxo cyclopentane 1-carboxylic acid, 3-oxo-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-oxo, 3-carboxycyclopentanone, cyclopentanone-3-carboxylic acid, 3-oxo-cyclopentanecarboxylic acid, 3-ketocyclopentanecarboxylic acid, 3-cyclopentanone-1 r-carboxylic acid |
PubChem CID | 227798 |
ChEBI | CHEBI:71213 |
IUPAC Name | 3-oxocyclopentane-1-carboxylic acid |
SMILES | OC(=O)C1CCC(=O)C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10071324
|
Thermo Scientific Acros
426340010 |
1 g | Glass bottle | ||||||
10081324
|
Thermo Scientific Acros
426340050 |
5 g | Glass bottle |
515.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
98-78-2 | |
128.13 | |
RDSNBKRWKBMPOP-UHFFFAOYNA-N | |
227798 | |
3-oxocyclopentane-1-carboxylic acid |
C6H8O3 | |
MFCD01320173 | |
3-oxocyclopentanecarboxylic acid, 3-oxo-cyclopentane-carboxylic acid, 3-oxo cyclopentane 1-carboxylic acid, 3-oxo-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-oxo, 3-carboxycyclopentanone, cyclopentanone-3-carboxylic acid, 3-oxo-cyclopentanecarboxylic acid, 3-ketocyclopentanecarboxylic acid, 3-cyclopentanone-1 r-carboxylic acid | |
CHEBI:71213 | |
OC(=O)C1CCC(=O)C1 |
Specifications
98-78-2 | |
100.0 | |
97% | |
MFCD01320173 | |
RDSNBKRWKBMPOP-UHFFFAOYNA-N | |
3-oxocyclopentane-1-carboxylic acid | |
227798 | |
128.13 | |
3-Oxo-1-cyclopentanecarboxylic acid |
96.0 | |
59°C to 63.5°C | |
C6H8O3 | |
3-oxocyclopentanecarboxylic acid, 3-oxo-cyclopentane-carboxylic acid, 3-oxo cyclopentane 1-carboxylic acid, 3-oxo-1-cyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-oxo, 3-carboxycyclopentanone, cyclopentanone-3-carboxylic acid, 3-oxo-cyclopentanecarboxylic acid, 3-ketocyclopentanecarboxylic acid, 3-cyclopentanone-1 r-carboxylic acid | |
OC(=O)C1CCC(=O)C1 | |
128.13 | |
CHEBI:71213 | |
97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
- 3-Oxo-1-cyclopentanecarboxylic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only