missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-3-(2-thienyl)propionitrile, 98%, Thermo Scientific Chemicals

Product Code. 11434293
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33898-90-7
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.18
MDL Number MFCD00052586
InChI Key XWWUQBHVRILEPB-UHFFFAOYSA-N
Synonym 2-thenoylacetonitrile, 3-oxo-3-thiophen-2-yl propanenitrile, 3-oxo-3-2-thienyl propanenitrile, 3-oxo-3-2-thienyl propionitrile, 2-thiophene-2-carbonyl acetonitrile, 3-oxo-3-thiophen-2-yl-propionitrile, 3-2-thienyl-3-oxopropanenitrile, pubchem8372, then-2-oylacetonitrile, acmc-1csfg
PubChem CID 141853
IUPAC Name 3-oxo-3-thiophen-2-ylpropanenitrile
SMILES O=C(CC#N)C1=CC=CS1

Specifications

Melting Point 132°C to 136°C
Quantity 5 g
UN Number UN3439
Beilstein 111990
Formula Weight 151.19
Percent Purity 98%
Chemical Name or Material 3-Oxo-3- (2-thienyl)propionitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.