Learn More
3-Phenoxyaniline, 98%, Thermo Scientific Chemicals
Description
3-Phenoxyaniline was used in the preparation of 4-phenoxyaniline-3?-bromopropionamide. It is also used in the design, synthesis and evaluation of potential inhibitors of HIV gp120-CD4 interactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Phenoxyaniline was used in the preparation of 4-phenoxyaniline-3′-bromopropionamide. It is also used in the design, synthesis and evaluation of potential inhibitors of HIV gp120-CD4 interactions.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Spezifikation
Spezifikation
Melting Point | 35°C to 37°C |
Density | 1.1583 |
Boiling Point | 329°C to 330°C |
Flash Point | >110°C (230°F) |
Molecular Formula | C6H10O3 |
Refractive Index | 1.1583 |
Quantity | 5 g |
Synonym | neopentylene carbonate, 2,2-dimethyltrimethylene carbonate, 5,5-dimethyl-1,3-diox-2-one, acmc-1adx1, 3-nitro-5-chlorobenzenesulfonamide, 5,5-dimethyltrimethylene carbonate, 5,5-dimethyl-1,3-dioxane-2-one, 5,5-dimethyl-2-oxo-1,3-dioxane, 1,3-dioxan-2-one,5,5-dimethyl, 5,5-dimethyl-1,3 dioxan-2-one |
Solubility Information | Slightly soluble in water. |
InChI Key | JRFXQKZEGILCCO-UHFFFAOYSA-N |
Mehr anzeigen |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10249068 | Certificate of Analysis | 09/02/2024 | A17495.14,A17495.06,A17495.22 |
10238807 | Certificate of Analysis | 24/02/2023 | A17495.14,A17495.06,A17495.22 |
10100150 | Certificate of Analysis | 23/11/2021 | A17495.14,A17495.06,A17495.22 |
J5873A | Certificate of Analysis | 23/11/2021 | A17495.14,A17495.06,A17495.22 |
10183246 | Certificate of Analysis | 23/11/2021 | A17495.14,A17495.06,A17495.22 |
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.