missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propyne, 97%, stabilized, Thermo Scientific Chemicals

Product Code. 10588643
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10147-11-2
Molecular Formula C9H8
Molecular Weight (g/mol) 116.16
MDL Number MFCD00134431
InChI Key NGKSKVYWPINGLI-UHFFFAOYSA-N
Synonym 3-phenyl-1-propyne, 2-propynyl benzene, benzylacetylene, prop-2-yn-1-ylbenzene, benzene, 2-propynyl, propargylbenzol, propargylbenzene, benzyl acetylene, 3-phenyl-propyne, 3-phenyl-1-propyn
PubChem CID 575753
IUPAC Name prop-2-ynylbenzene
SMILES C#CCC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9340g/mL
Boiling Point 75.0°C (20.0 mmHg)
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5260 to 1.5290
Quantity 5 g
Specific Gravity 0.934
Stabilizer 250ppm BHT
Formula Weight 116.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Phenyl-1-propyne, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Phenyl-1-propyne, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.