missing translation for 'onlineSavingsMsg'
Learn More

3-(Phenylamino)propyltrimethoxysilane, 96%, Thermo Scientific Chemicals

Product Code. 11343796
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3068-76-6
Molecular Formula C12H21NO3Si
Molecular Weight (g/mol) 255.389
MDL Number MFCD00053673
InChI Key KBJFYLLAMSZSOG-UHFFFAOYSA-N
Synonym n-3-trimethoxysilyl propyl aniline, trimethoxy 3-phenylamino propyl silane, n-phenylaminopropyltrimethoxysilane, unii-7au1h1fv6a, benzenamine, n-3-trimethoxysilyl propyl, n-phenyl-3-aminopropyltrimethoxysilane, 7au1h1fv6a, n-3-trimethoxysilylpropyl aniline, 3-phenylamino propyltrimethoxysilane
PubChem CID 76476
IUPAC Name N-(3-trimethoxysilylpropyl)aniline
SMILES CO[Si](CCCNC1=CC=CC=C1)(OC)OC

Specifications

Boiling Point 310°C
Quantity 25 g
Linear Formula C6H5NH(CH2)3Si(OCH3)3
UN Number UN3267
Beilstein 8620045
Flash Point 165°C (329°F)
Formula Weight 255.39
Percent Purity 96%
Odor Weak
Refractive Index 1.5055
Sensitivity Moisture sensitive
Density 1.07
Chemical Name or Material 3-(Phenylamino)propyltrimethoxysilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'