Learn More
3-Phenylpropionic acid, 99%, Thermo Scientific Chemicals
Description
Hydrocinnamic acid is inhibitor of carboxypeptidase. Hydrocinnamic acid was used in the synthesis of three medium-chain acyl-Coenzyme A?s i.e. 3-phenylpropionyl-CoA from mixed anhydrides of fatty acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Hydrocinnamic acid is inhibitor of carboxypeptidase. Hydrocinnamic acid was used in the synthesis of three medium-chain acyl-Coenzyme A′s i.e. 3-phenylpropionyl-CoA from mixed anhydrides of fatty acids.
Solubility
Soluble in water
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | 47°C to 50°C |
Density | 1.071 |
Boiling Point | 280°C |
Flash Point | >110°C (230°F) |
Quantity | 100 g |
Beilstein | 907515 |
Merck Index | 14,4784 |
Solubility Information | Soluble in water |
Formula Weight | 150.18 |
Percent Purity | 99% |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M09J029 | Certificate of Analysis | 26/01/2023 | A14908.22,A14908.36 |
5001G14R | Certificate of Analysis | 22/11/2021 | A14908.22,A14908.36 |
10179550 | Certificate of Analysis | 22/11/2021 | A14908.22,A14908.36 |
F7611A | Certificate of Analysis | 22/11/2021 | A14908.22,A14908.36 |
10149783 | Certificate of Analysis | 22/11/2021 | A14908.22,A14908.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.