missing translation for 'onlineSavingsMsg'
Learn More

3-Picoline, 99%

Product Code. 10347110 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
500mL
This item is not returnable. View return policy

Product Code. 10347110

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-99-6 | C6H7N | 93.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-99-6
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00006402
InChI Key ITQTTZVARXURQS-UHFFFAOYSA-N
Synonym 3-picoline, beta-picoline, pyridine, 3-methyl, m-picoline, m-methylpyridine, b-picoline, beta-methylpyridine, .beta.-methylpyridine, .beta.-picoline, meta-methylpyridine
PubChem CID 7970
ChEBI CHEBI:39922
IUPAC Name 3-methylpyridine
SMILES CC1=CN=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -19.0°C
Color Yellow
Density 0.9500g/mL
Boiling Point 143.0°C to 144.0°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5050 to 1.5070
Quantity 500 mL
Beilstein 20, 239
Merck Index 15, 7513
Specific Gravity 0.95
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 93.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Picoline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.