missing translation for 'onlineSavingsMsg'
Learn More

3-Picoline-N-oxide, 98%, Thermo Scientific™

Product Code. 10081310
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1003-73-2
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006203
InChI Key DMGGLIWGZFZLIY-UHFFFAOYSA-N
Synonym 3-methylpyridine 1-oxide, 3-picoline-n-oxide, 3-picoline n-oxide, 3-methylpyridine n-oxide, 3-methylpyridine-n-oxide, pyridine, 3-methyl-, 1-oxide, 3-picoline 1-oxide, beta-picoline n-oxide, 3-methylpyridin-1-ium-1-olate, 3-methylpyridine-1-oxide
PubChem CID 13858
IUPAC Name 3-methyl-1-oxidopyridin-1-ium
SMILES CC1=C[N+](=CC=C1)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 38°C to 40°C
Color Brown to Yellow
Boiling Point 150°C (15.0 mmHg)
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: freely soluble in water
Formula Weight 109.13
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 3-Picoline-N-oxide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Picoline-N-oxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.