Learn More
3-Piperidin-1-ylmethyl benzylamine, ≥97%, Thermo Scientific™
101.00 € - 341.00 €
Chemical Identifiers
CAS | 91271-80-6 |
---|---|
Molecular Formula | C13H20N2 |
Molecular Weight (g/mol) | 204.317 |
MDL Number | MFCD06408781 |
InChI Key | BMNFPARGJHNMAX-UHFFFAOYSA-N |
Synonym | 3-piperidin-1-ylmethyl-benzylamine, 1-3-piperidin-1-ylmethyl phenyl methanamine, 3-piperidin-1-ylmethyl phenyl methanamine, 3-piperidin-1-ylmethyl benzyl amine, 3-piperidylmethyl phenyl methylamine, 3-piperidin-1-ylmethylbenzylamine, benzenemethanamine, 3-1-piperidinylmethyl, 3-piperidin-1-yl methyl phenyl methanamine, 1-3-piperidin-1-ylmethyl phenyl methylamine |
PubChem CID | 2794678 |
IUPAC Name | [3-(piperidin-1-ylmethyl)phenyl]methanamine |
SMILES | C1CCN(CC1)CC2=CC(=CC=C2)CN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10513423
|
Thermo Scientific Maybridge
MO01157CB |
250 mg |
101.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10219932
|
Thermo Scientific Maybridge
MO01157DA |
1 g |
262.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10081004
|
Thermo Scientific Maybridge
MO01157DE |
5 g |
341.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
91271-80-6 | |
204.317 | |
BMNFPARGJHNMAX-UHFFFAOYSA-N | |
2794678 | |
C1CCN(CC1)CC2=CC(=CC=C2)CN |
C13H20N2 | |
MFCD06408781 | |
3-piperidin-1-ylmethyl-benzylamine, 1-3-piperidin-1-ylmethyl phenyl methanamine, 3-piperidin-1-ylmethyl phenyl methanamine, 3-piperidin-1-ylmethyl benzyl amine, 3-piperidylmethyl phenyl methylamine, 3-piperidin-1-ylmethylbenzylamine, benzenemethanamine, 3-1-piperidinylmethyl, 3-piperidin-1-yl methyl phenyl methanamine, 1-3-piperidin-1-ylmethyl phenyl methylamine | |
[3-(piperidin-1-ylmethyl)phenyl]methanamine |
Specifications
91271-80-6 | |
97% | |
C13H20N2 | |
3-piperidin-1-ylmethyl-benzylamine, 1-3-piperidin-1-ylmethyl phenyl methanamine, 3-piperidin-1-ylmethyl phenyl methanamine, 3-piperidin-1-ylmethyl benzyl amine, 3-piperidylmethyl phenyl methylamine, 3-piperidin-1-ylmethylbenzylamine, benzenemethanamine, 3-1-piperidinylmethyl, 3-piperidin-1-yl methyl phenyl methanamine, 1-3-piperidin-1-ylmethyl phenyl methylamine | |
C1CCN(CC1)CC2=CC(=CC=C2)CN | |
204.317 | |
204.32 | |
3-Piperidin-1-ylmethyl benzylamine |
97.0 | |
Amber glass bottle | |
MFCD06408781 | |
BMNFPARGJHNMAX-UHFFFAOYSA-N | |
[3-(piperidin-1-ylmethyl)phenyl]methanamine | |
2794678 | |
97% |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica
GHS Signal Word: Danger