missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinobenzaldehyde, 95%, Thermo Scientific™

Product Code. 10133724
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 669050-72-0
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
InChI Key FXXQUTSXXILOMY-UHFFFAOYSA-N
Synonym 3-piperidinobenzaldehyde, 3-piperidin-1-yl benzaldehyde, benzaldehyde, 3-1-piperidinyl, 3-piperidylbenzaldehyde, 3-1-piperidinyl benzaldehyde
PubChem CID 7164587
IUPAC Name 3-piperidin-1-ylbenzaldehyde
SMILES C1CCN(CC1)C2=CC=CC(=C2)C=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 189.26
Percent Purity 95%
Chemical Name or Material 3-Piperidinobenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'