missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinobenzaldehyde, 95%, Thermo Scientific™

Product Code. 10452083
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 669050-72-0
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
InChI Key FXXQUTSXXILOMY-UHFFFAOYSA-N
Synonym 3-piperidinobenzaldehyde, 3-piperidin-1-yl benzaldehyde, benzaldehyde, 3-1-piperidinyl, 3-piperidylbenzaldehyde, 3-1-piperidinyl benzaldehyde
PubChem CID 7164587
IUPAC Name 3-piperidin-1-ylbenzaldehyde
SMILES C1CCN(CC1)C2=CC=CC(=C2)C=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 189.26
Percent Purity 95%
Chemical Name or Material 3-Piperidinobenzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.