missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Piperidinobenzaldehyde, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 189.26 |
Percent Purity | 95% |
Chemical Name or Material | 3-Piperidinobenzaldehyde |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction