missing translation for 'onlineSavingsMsg'
Learn More

[3-(Piperidinomethyl)phenyl]methanol, ≥90%, Thermo Scientific™

Product Code. 10095952
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 73278-91-8
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C13H19NO
MDL Number MFCD07772858
Quantity 1 g
Synonym 3-piperidinomethyl phenyl methanol, 3-piperidin-1-ylmethyl phenyl methanol, 3-piperidinomethylbenzylalcohol, 3-1-piperidylmethyl-benzyl alcohol, 3-1-piperidinylmethyl benzenemethanol, 3-piperidylmethyl phenyl methan-1-ol
InChI Key NZKXLHGCZWDXGE-UHFFFAOYSA-N
SMILES C1CCN(CC1)CC2=CC(=CC=C2)CO
IUPAC Name [3-(piperidin-1-ylmethyl)phenyl]methanol
Molecular Weight (g/mol) 205.301
PubChem CID 7164645
Formula Weight 205.3
Percent Purity 90%
Chemical Name or Material [3-(Piperidinomethyl)phenyl]methanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.