Learn More
Fisherbrand™ 3-Ply Sterile Facemasks
3-Ply Sterile Facemasks constructed of polyethylene apertured film and PTFE (polytetrafluoroethylene).
Brand:  Fisherbrand™ 12-891-00217
 View more versions of this product
Product Code. 17397289
  • 167.00 € / Pack of 200
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Constructed from a combination of pleated clear, polyethylene apertured film and PTFE (polytetrafluoroethylene), with soft tie strings
  • Manufactured in systems complying ISO 13485:2016 requirements
  • Certified Sterile
Specifications
Specifications
Cleanroom Face Mask
Sterile
The masks are rigorously tested and manufactured in an ISO 13485 certified environment. The tests are validated by an independent control laboratory. This product is not a medical device in context of Regulation (EU) 2017/745/Directive 93/42/EEC nor personal protective equipment in context of Regulation (EU) 2016/425. Each batch is sterilized following ISO 11137 and substantiate a sterility assurance level SAL 10−6 according to ISO 11137-2:2013.
Industrial Use Only
White
Pleated with Ties
Single-use
17 cm