missing translation for 'onlineSavingsMsg'
Learn More

3-(Pyridin-2-yloxy)aniline, 97%, Thermo Scientific™

Product Code. 10235964
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 86556-09-4
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.21
MDL Number MFCD06825506
InChI Key CLQMOPKXIBQKKG-UHFFFAOYSA-N
Synonym 3-pyridin-2-yloxy aniline, 3-pyridin-2-yloxy-phenylamine, 3-2-pyridyloxy aniline, 3-2-pyridyloxy phenylamine, 3-pyridin-2-yl oxy aniline, benzenamine,3-2-pyridinyloxy, benzenamine, 3-2-pyridinyloxy
PubChem CID 13150573
IUPAC Name 3-pyridin-2-yloxyaniline
SMILES NC1=CC(OC2=CC=CC=N2)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 3-(Pyridin-2-yloxy)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'