missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-5-ylaniline, ≥97%, Thermo Scientific™

Product Code. 10558253
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 69491-59-4
Molecular Formula C10H9N3
Molecular Weight (g/mol) 171.203
MDL Number MFCD06802522
InChI Key DZEIKJMNXHOFHL-UHFFFAOYSA-N
PubChem CID 7162049
IUPAC Name 3-pyrimidin-5-ylaniline
SMILES C1=CC(=CC(=C1)N)C2=CN=CN=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 3-pyrimidin-5-yl aniline, 3-5-pyrimidinyl aniline, benzenamine,3-5-pyrimidinyl, 3-5-pyrimidyl aniline, 3-5-pyrimidyl-aniline, 3-pyrimidin-5-ylphenylamine, 3-pyrimidin-5-yl-phenylamine, 3-pyrimidin-5-yl phenylamine, benzenamine, 3-5-pyrimidinyl
Formula Weight 171.2
Percent Purity 97%
Chemical Name or Material 3-Pyrimidin-5-ylaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'