missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ 3-Stop Tubing, Tygon LMT-55

Designed for optimal operation in your Ismatec pump.

Brand:  Masterflex™ 97619-42

 View more versions of this product

Product Code. 15945811

  • 186.00 € / Pack of 12
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with pumps that incorporate cassettes into the pumphead design.
  • Color-coded stops.
  • Inexpensive all-around tubing for general laboratory applications.
  • Transparent.
  • Resistant to almost all inorganic chemicals.
  • Smooth polished inner wall.
  • Low gas permeability.
  • Non-aging and non-oxidizing.
Specifications

Specifications

Purple-Purple
Tygon LMT-55
General laboratory applications, pumps with cassettes in pumphead design
Acids: Good, Alkaline solutions: Good, Solvents: Not rec., Pressure: Fair, Vacuum: Good, Viscous media: Excellent, Sterile media: Limited
Transparent
2.06 mm
406.4 mm
12/Pk
I.D. 2.06 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.