missing translation for 'onlineSavingsMsg'
Learn More
3-Sulfobenzoic acid disodium salt monohydrate, 97%, Thermo Scientific™ Product Code.: 11359829

3-Sulfobenzoic acid disodium salt monohydrate, 97%, Thermo Scientific™

Product Code. 11359829
25 g
Click to view available options
Quantity:
25 g
500 g
This item is not returnable. View return policy

Product Code. 11359829

Brand: Thermo Scientific Alfa Aesar B24154.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14995-40-5
Molecular Formula C7H4O5S
Molecular Weight (g/mol) 200.17
MDL Number MFCD00149379
InChI Key QMWGSOMVXSRXQX-UHFFFAOYSA-L
Synonym sodium 3-sulfonatobenzoate hydrate, 3-sulfobenzoic acid disodium salt monohydrate, disodium 3-sulfonatobenzoate hydrate
PubChem CID 2734925
SMILES [O-]C(=O)C1=CC=CC(=C1)S([O-])(=O)=O

Specifications

Quantity 25 g
IUPAC Name disodium;3-sulfonatobenzoate;hydrate
Formula Weight 264.17 (246.15 Anhydrous)
Percent Purity 97%
Chemical Name or Material 3-Sulfobenzoic acid disodium salt monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Sulfobenzoic acid disodium salt monohydrate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.