missing translation for 'onlineSavingsMsg'
Learn More
3-Sulfolene, 98%, Thermo Scientific Chemicals Product Code.: 11319459

3-Sulfolene, 98%, Thermo Scientific Chemicals

Product Code. 11319459
500 g
Click to view available options
Quantity:
100 g
500 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11319459

Brand: Thermo Scientific Alfa Aesar A13887.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is a source of cisoid butadiene for Diels-Alder reactions. This chemical is also a convenient source of sulfur dioxide. It is a solvent used in the petrochemical industry for the extraction of aromatics from hydrocarbon streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a source of cisoid butadiene for Diels-Alder reactions. This chemical is also a convenient source of sulfur dioxide. It is a solvent used in the petrochemical industry for the extraction of aromatics from hydrocarbon streams.

Solubility
Soluble in water, ethanol, benzene, ether.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-79-2
Molecular Formula C4H6O2S
Molecular Weight (g/mol) 118.15
MDL Number MFCD00005481
InChI Key MBDNRNMVTZADMQ-UHFFFAOYSA-N
Synonym 3-sulfolene, butadiene sulfone, sulfolene, thiophene, 2,5-dihydro-, 1,1-dioxide, beta-sulfolene, 2,5-dihydrothiophene sulfone, 2,5-dihydrothiophene dioxide, 2,5-dihydrothiophene-1,1-dioxide, sulfol-3-ene, ccris 569
PubChem CID 6498
IUPAC Name 2,5-dihydrothiophene 1,1-dioxide
SMILES C1C=CCS1(=O)=O

Specifications

Melting Point 63°C to 67°C
Density 1.314
Flash Point 112°C (233°F)
Quantity 500 g
Beilstein 107004
Merck Index 14,8956
Solubility Information Soluble in water,ethanol,benzene,ether.
Formula Weight 118.15
Percent Purity 98%
Chemical Name or Material 3-Sulfolene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465870 Certificate TypeCertificate of Analysis Date17/09/2024 Catalog NumberA13887.0I,A13887.36,A13887.0B,A13887.22
Lot NumberA0462051 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA13887.0I,A13887.36,A13887.0B,A13887.22
Lot NumberA0462049 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA13887.0I,A13887.36,A13887.0B,A13887.22
Lot NumberA0459905 Certificate TypeCertificate of Analysis Date12/03/2024 Catalog NumberA13887.0I,A13887.36,A13887.0B,A13887.22
Lot Number10240682 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog NumberA13887.0I,A13887.36,A13887.0B,A13887.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Sulfolene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.