missing translation for 'onlineSavingsMsg'
Learn More

3-Thienylmethylamine, 97%, Thermo Scientific™

Product Code. 10394322
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 27757-86-4
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.178
MDL Number MFCD01529872
InChI Key DUDAKCCDHRNMDJ-UHFFFAOYSA-N
Synonym 3-thienylmethylamine, 3-aminomethyl thiophene, 3-thiophenemethanamine, 3-thiophenemethylamine, 1-thien-3-ylmethanamine, 3-thienylmethyl amine, 1-3-thienyl methanamine, thiophen-3-yl-methylamine, thiophen-3-yl methanamine, thien-3-yl methylamine
PubChem CID 2776381
IUPAC Name thiophen-3-ylmethanamine
SMILES C1=CSC=C1CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 113.18
Percent Purity 97%
Chemical Name or Material 3-Thienylmethylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'