missing translation for 'onlineSavingsMsg'
Learn More

3-Tolylboronic acid, 97%, Thermo Scientific Chemicals

29.90 € - 53.50 €

Chemical Identifiers

CAS 17933-03-8
Molecular Formula C7H9BO2
Molecular Weight (g/mol) 135.96
MDL Number MFCD00040198
InChI Key BJQCPCFFYBKRLM-UHFFFAOYSA-N
Synonym 3-tolylboronic acid, m-tolylboronic acid, 3-methylphenyl boronic acid, 3-methylbenzeneboronic acid, 3-methyl phenyl boronic acid, 3-tolyboronic acid, 3-methylphenyl boranediol, m-methylphenylboronic acid, m-tolyl boronic acid
PubChem CID 2733950
IUPAC Name (3-methylphenyl)boronic acid
SMILES CC1=CC=CC(=C1)B(O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10778362
View Documents Promotion Details
Thermo Scientific Acros
344420010
1 g Glass bottle
29.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
10778552
View Documents Promotion Details
Thermo Scientific Acros
344420050
5 g Glass bottle
53.50 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17933-03-8
135.96
BJQCPCFFYBKRLM-UHFFFAOYSA-N
2733950
CC1=CC=CC(=C1)B(O)O
C7H9BO2
MFCD00040198
3-tolylboronic acid, m-tolylboronic acid, 3-methylphenyl boronic acid, 3-methylbenzeneboronic acid, 3-methyl phenyl boronic acid, 3-tolyboronic acid, 3-methylphenyl boranediol, m-methylphenylboronic acid, m-tolyl boronic acid
(3-methylphenyl)boronic acid

Specifications

17933-03-8
100.0
Beige
97%
CH3C6H4B(OH)2
3-tolylboronic acid, m-tolylboronic acid, 3-methylphenyl boronic acid, 3-methylbenzeneboronic acid, 3-methyl phenyl boronic acid, 3-tolyboronic acid, 3-methylphenyl boranediol, m-methylphenylboronic acid, m-tolyl boronic acid
BJQCPCFFYBKRLM-UHFFFAOYSA-N
(3-methylphenyl)boronic acid
2733950
97%
3-Tolylboronic acid, 97%
96.0
160.0°C to 162.0°C
Authentic
C7H9BO2
MFCD00040198
Solubility in water: +/- 2.5% in water.
CC1=CC=CC(=C1)B(O)O
135.96
135.96
Chunks and/or Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

missing translation for 'tsca' : TSCA

Product Identifier
  • 3-Tolylboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only