missing translation for 'onlineSavingsMsg'
Learn More

3'-(Trifluoromethoxy)acetophenone, 97%, Thermo Scientific Chemicals

Artikelnummer. 11413387
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 170141-63-6
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD00042403
InChI Key UYHTUQHYGKAYJM-UHFFFAOYSA-N
Synonym 3'-trifluoromethoxy acetophenone, 1-3-trifluoromethoxy phenyl ethanone, 3-trifluoromethoxy acetophenone, 1-3-trifluoromethoxy phenyl ethan-1-one, 3-trifluoromethoxyacetophenone, 1-acetyl-3-trifluoromethoxy benzene, fxffor cv1, pubchem4297, intermediates-zcf02268
PubChem CID 737161
IUPAC Name 1-[3-(trifluoromethoxy)phenyl]ethanone
SMILES CC(=O)C1=CC=CC(OC(F)(F)F)=C1

Spezifikation

Density 1.285
Boiling Point 49°C to 51°C (13 mmHg)
Flash Point 83°C (181°F)
Refractive Index 1.4515
Quantity 1 g
Beilstein 6196333
Formula Weight 204.15
Percent Purity 97%
Chemical Name or Material 3'-(Trifluoromethoxy)acetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'