missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)phenyltrimethylammonium hydroxide, 5% w/v in methanol, Thermo Scientific Chemicals

Product Code. 11359408
Click to view available options
Quantity:
5 mL
25 mL

Transesterification reactions

Transesterification reactions.3-(Trifluoromethyl)phenyltrimethylammonium hydroxide is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68254-41-1
Molecular Formula C10H14F3NO
Molecular Weight (g/mol) 221.223
MDL Number MFCD00059473
InChI Key BFPOZPZYPNVMHU-UHFFFAOYSA-M
Synonym 3-trifluoromethyl phenyltrimethylammonium hydroxide, m-tfptah, trifluoromethylphenyltrimethylammoniumhydroxide, 3-trifluoromethyl phenyltrimethylammonium hydroxide in methanol, n,n,n-trimethyl-3-trifluoromethyl anilinium hydroxide, trimethyl-3-trifluoromethyl phenyl azanium hydroxide, m-trifluoromethylphenyl trimethylammonium hydroxide, trimethyl-3-trifluoromethylphenylammonium hydroxide, n,n,n-trimethyl-3-trifluoromethyl anilinium oxidanide, trimethyl-alpha,alpha,alpha-trifluoro-m-tolylammonium hydroxide
PubChem CID 2760772
IUPAC Name trimethyl-[3-(trifluoromethyl)phenyl]azanium;hydroxide
SMILES C[N+](C)(C)C1=CC=CC(=C1)C(F)(F)F.[OH-]

Specifications

Quantity 5 mL
UN Number UN2924
Solubility Information Miscible with water and alcohols.
Formula Weight 221.22
Concentration or Composition (by Analyte or Components) 5% w/v in methanol
Physical Form Liquid
Chemical Name or Material 3- (Trifluoromethyl)phenyltrimethylammonium hydroxide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-(Trifluoromethyl)phenyltrimethylammonium hydroxide, 5% w/v in methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Methanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.