missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethylthio)benzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 11408857
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 946-65-6
Molecular Formula C8H4F3O2S
Molecular Weight (g/mol) 221.17
MDL Number MFCD00236337
InChI Key IVGAPIVNQABETQ-UHFFFAOYSA-M
Synonym 3-trifluoromethylthio benzoic acid, 3-trifluoromethyl thio benzoic acid, 3-trifluoromethyl sulfanyl benzoic acid, 3-trifluoromethylsulfanyl benzoic acid, 3-trifluoromethylsulfanyl-benzoic acid, benzoic acid, 3-trifluoromethyl thio, 3-trifluoromethyl sulphanyl benzoic acid, 3-trifluoromethylthio benzoicacid, 3-carboxyphenyl trifluoromethyl sulphide
PubChem CID 688330
IUPAC Name 3-(trifluoromethylsulfanyl)benzoic acid
SMILES [O-]C(=O)C1=CC=CC(SC(F)(F)F)=C1

Specifications

Melting Point 71°C to 74°C
Quantity 1 g
Formula Weight 222.18
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethylthio)benzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'