missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethoxysilyl)propyl acrylate, 90%, stabilized, Thermo Scientific Chemicals

Product Code. 10716505
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4369-14-6
Molecular Formula C9H18O5Si
Molecular Weight (g/mol) 234.32
MDL Number MFCD00054803
InChI Key KBQVDAIIQCXKPI-UHFFFAOYSA-N
Synonym 3-trimethoxysilyl propyl acrylate, 3-acryloyloxy propyltrimethoxysilane, 3-acryloxypropyltrimethoxysilane, 2-propenoic acid, 3-trimethoxysilyl propyl ester, 3-acryloxypropyl trimethoxysilane, 3-trimethoxysilyl propyl 2-propenoate, 3-acryloyloxypropyltrimethoxysilane, 3-acryloxypropyltrimethoxysilane, stab. with 100ppm bht, acmc-1aoz6, acrylic acid, 3-trimethoxysilyl propyl ester
PubChem CID 151204
IUPAC Name 3-trimethoxysilylpropyl prop-2-enoate
SMILES CO[Si](CCCOC(=O)C=C)(OC)OC

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Colorless
Density 1.0590g/mL
Boiling Point 95°C (3.0 mmHg)
Flash Point 123°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCO2(CH2)3Si(OCH3)3
Refractive Index 1.4280 to 1.4300 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.059
Stabilizer stabilized with BHT
Formula Weight 234.32
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 3-(Trimethoxysilyl)propyl acrylate, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-(Trimethoxysilyl)propyl acrylate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'