missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethoxysilyl)propyl methacrylate, 98%, Thermo Scientific Chemicals

Product Code. 10412411
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2530-85-0
Molecular Formula C10H20O5Si
Molecular Weight (g/mol) 248.35
MDL Number MFCD00008593
InChI Key XDLMVUHYZWKMMD-UHFFFAOYSA-N
Synonym 3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester
PubChem CID 17318
IUPAC Name 3-trimethoxysilylpropyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCC[Si](OC)(OC)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.0400g/mL
Boiling Point 190.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2(CH2)3Si(OCH3)3
Refractive Index 1.4300 to 1.4320
Quantity 5 mL
Specific Gravity 1.04
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,ether,methanol and,hydrocarbons
Viscosity 2.5 mm2/s (25°C)
Formula Weight 248.35
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-(Trimethoxysilyl)propyl methacrylate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.