missing translation for 'onlineSavingsMsg'
Learn More

3-[(Trimethylsilyl)ethynyl]benzonitrile, 97%, Thermo Scientific Chemicals

Product Code. 15441128
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 190771-22-3
Molecular Formula C12H13NSi
Molecular Weight (g/mol) 199.328
MDL Number MFCD09260494
InChI Key QEZDMZVLNWSJSA-UHFFFAOYSA-N
Synonym 3-trimethylsilyl ethynyl benzonitrile, 3-2-trimethylsilyl ethynyl benzonitrile, benzonitrile,3-2-trimethylsilyl ethynyl, benzonitrile, 3-2-trimethylsilyl ethynyl, 3-trimethylsilanylethynylbenzonitrile, 3-trimethylsilanylethynyl-benzonitrile
PubChem CID 22570318
IUPAC Name 3-(2-trimethylsilylethynyl)benzonitrile
SMILES C[Si](C)(C)C#CC1=CC(=CC=C1)C#N

Specifications

Melting Point 42°C to 47°C
Quantity 250 mg
Formula Weight 199.33
Percent Purity 97%
Chemical Name or Material 3-[(Trimethylsilyl)ethynyl]benzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.