missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 3 Way Test Strip

With a chlorine range of 0 to 20 mg/L and a pH or 6 to 9, these 3-in-1 Pooltest test strips are ideal for estimating basic water chemistry.

Brand:  Fisherbrand™ PT634

Product Code. 11517733

  • 23.32 € / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

    • Test strips, 3 in 1, Pooltest
    • For estimating basic water chemistry
    • 1 set test kit, 3 in 1, 50 tests, chlorine range: 0 to 20mg/L, pH range: 6 to 9, alkalinity
    • Test strips, bromine, pH and alkalinity, 50 tests
Specifications

Specifications

6 to 9
1 Each
3 Way Test Strip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.