Learn More
Gilson™ 3-Way Valve Kit
These assemblies and packages add fraction collection capabilities to various Gilson instruments.
Brand:  Gilson™ 170730
 View more versions of this product
Product Code. 16686135
  • 1380.00 € / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Purification Technique HPLC
  • Used With Fraction Collector
Specifications
Specifications
FC203
1.5 mm