missing translation for 'onlineSavingsMsg'
Learn More

Econix™ Bio-bins™ Single M Stand for 30 L Bio-bin Waste Container - Stainless Steel

Use Econix™ Stainless Steel 'M' stand designed to allow the 30 L Bio-bin™ to tilt at approximately 15 degrees for easy filling of tall thin items like pipettes and other general non-sharps laboratory waste.

Brand:  Econix™ BEXBBMS30.0

Product Code. 15817221

  • 74.30 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Description

Description

  • For use with 30L Bio-bin™
    • Allows the user to see the contents easily
    • Helps to prevent over filling
    • For use within laboratories and next to work benches
Specifications

Specifications

400 mm
Autoclavable
Stainless Steel
Stainless Steel
30 L Bio-bin™
Stand
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.