missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ 300mL Enviro Dipper

A simple yet effective method of taking a surface sample.

Brand:  Sampling Systems™ A725-300

Product Code. 15879711

  • 12.40 € / Each
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Simply lower the bucket into the liquid
  • Manufactured from stainless steel
  • Can be ordered with either a stainless steel chain or a braided polypropylene cord
  • Holds 300mL
Specifications

Specifications

Sampling Bucket
300 mL
Stainless Steel
Metallic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.