missing translation for 'onlineSavingsMsg'
Learn More
Cole-Parmer™ 304 Stainless Steel Cover Product Code.: 11747974
Estimated Shipment: 20-06-2025
to see stock.
Add to basket

Cole-Parmer™ 304 Stainless Steel Cover

Product Code. 11747974 Shop All Cole-Parmer Products
240mm
Click to view available options
Diameter (Metric) Outer:
171.45 mm
222mm
240mm
280mm
This item is not returnable. View return policy

Product Code. 11747974

Brand: Cole-Parmer™ 0720710

Estimated Shipment: 20-06-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

For use with stainless steel beakers

  • Covers reduce evaporation
  • Contain harmful vapors, or keep contaminants out

Specifications

Diameter (Metric) Outer 240mm
Material Stainless Steel (304)
Quantity 1Each
Certifications/Compliance AISI 304
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ 304 Stainless Steel Cover >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.