missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Stainless Steel Offset Spoons
Description
- Laboratory Offset Spoon
- Stainless steel 304 spec 18/8 non-magnetic, bright finish
- Uses the finest in corrosion-resistant 304 stainless steel
- The unique curved shape spoon, offset, allows contact with all corners of bottles and containers reducing sample losses
- The alternate end can be used as a scraper to chip away at hard samples or used as a spatula for sampling or lifting
- Can be used within laboratory to retrieve powders, granules, solids and viscous liquids
Specifications
1 Ea. | |
Autoclavable | |
Bright | |
-196°C to +550°C | |
190 mm |
Sterile | |
Laboratory Offset Spoon | |
For Sterile Applications | |
Stainless Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction