Learn More
Sampling Systems™ 304 Stainless Steel Storage Containers without Toggle Clamps
These 304 Stainless Steel Storage Containers are an excellent option for multi-purpose storage in the laboratory.
Brand:  Sampling Systems™ A454-30
 View more versions of this product
Product Code. 15819681
  • 845.00 € / Each
Explore more special offers
Description
Description
  • Manufactured from durable 304 grade stainless steel
  • Crevice-free interior
  • Close fitting recessed lid
  • Each with two sturdy handles
  • Flat bottom with chime ring to give added protection
  • Light duty construction
  • Available in a wide range of capacities to fit your specific needs
Specifications
Specifications
Yes
Recessed Lid with Handle
Multi-purpose Storage
445 mm
Storage container
30 L
Stainless Steel
300 mm