Learn More
Thermo Scientific™ 316 Stainless Steel Capillary Tubing for HPLC in 5-Foot Coils
Use cleaned, polished and passivated Thermo Scientific™ 316 Stainless Steel Capillary Tubing for HPLC, ready for immediate use.
Brand:  Thermo Scientific™ U-106
 View more versions of this product
Product Code. 10488294
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Alert:
Not recommended for biological samples. Rough internal surface may lead to sample carryover. Reasonably flexible, but works best with minimal bending
Description
Description
Use Thermo Scientific™ 316 Stainless Steel Capillary Tubing for ultrahigh-pressure applications. Prefinished, square, burr-free ends and interiors ensure minimum dead volume connections. Cleaned, polished and passivated, it is ready for immediate use. This tubing is not recommended for biological samples.
  • Suitable for ultra high pressure applications
  • Wide chemical compatibilty
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HPLC
0.010 in. ID; 5 ft. length
0.010 in.
5 ft.
Stainless Steel
Capillary Tubing
HPLC
0.25 mm
1.524 m
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificats
Promotions
Promotions
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.