missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Tubes™ 3810X
GREENER_CHOICE

The easy-to-open, securely closing, Eppendorf Tubes™ 3810X offers comfort and dependability for sample preparation, centrifugation, and storage. One vessel for all applications in molecular biology, chemistry, and biochemistry. Safer than ever: designed for centrifugation up to 30,000 x g.

49.36 € - 77.81 €

Specifications

Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Material Polypropylene
No. per Pack 1000 Pk.
Temperature Range (English) -122.8°F to +212°F
View More Specs

Products 6
Product Code Brand Color Purity or Quality Grade Price Quantity & Availability  
Product Code Brand Color Purity or Quality Grade Price Quantity & Availability  
10154671
GREENER_CHOICE
View Documents
Eppendorf™
0030125215
Clear PCR Clean
50.95 €
Pack of 1000
Estimated Shipment: 06-08-2025
to see stock.
Add to basket
 
10451043
GREENER_CHOICE
View Documents
Eppendorf™
0030125150
Clear Eppendorf Quality
49.36 €
Pack of 1000
In Stock
to see stock.
Edge
Add to basket
 
10758052
GREENER_CHOICE
View Documents
Eppendorf™
0030125185
Green Eppendorf Quality
77.81 €
Pack of 1000
In Stock
to see stock.
Add to basket
 
10277391
GREENER_CHOICE
View Documents
Eppendorf™
0030125177
Blue Eppendorf Quality
74.65 €
Pack of 1000
In Stock
to see stock.
Add to basket
 
10596901
GREENER_CHOICE
View Documents
Eppendorf™
0030125207
Yellow Eppendorf Quality
74.65 €
Pack of 1000
In Stock
to see stock.
Add to basket
 
10649781
GREENER_CHOICE
View Documents
Eppendorf™
0030125193
Red Eppendorf Quality
77.81 €
Pack of 1000
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Use these tubes for all applications in molecular biology, clinical chemistry and biochemistry. Eppendorf™ 3810X Polypropylene Microcentrifuge Tube offers comfort and dependability for sample preparation, centrifugation and storage.

  • Tube lid is easy to open and close for ergonomic operation.
  • Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes.
  • g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples.
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
  • Graduation for visual control of the pipetted volume.
  • Frosted lid and surface on the side for easy labeling.
  • Ensured functionality from -86°C to 100°C.
  • Autoclavable when open (121°C, 20 min).
  • Available in Eppendorf Quality and lot-certified PCR clean purity grade.
Specifications

Specifications

Autoclavable
Polypropylene
-122.8°F to +212°F
Non-sterile
0.05 oz.
Cylindrical
30,000 x g
1.5 mL
1000 Pk.
-86°C to +100°C
Yes
Flat Frost Lid
Conical
Microcentrifuge Tube
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Tubes™ 3810X

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.