Learn More
3BDO, MedChemExpress
MedChemExpress 3BDO is a new mTOR activator which can also inhibit autophagy.
Brand:  Medchem Express HY-U00434
Encompass_Preferred
 View more versions of this product
Product Code. 16404106
  • 368.55 € / 25mg
Explore more special offers
Description
Description
3BDO is a new mTOR activator which can also inhibit autophagy.
Specifications
Specifications
3BDO
Light Yellow
C18H17NO5
DMSO : ≥ 100 mg/mL (305.50 mM) ∣Ethanol : 20 mg/mL (61.10 mM; Need ultrasonic)
327.33
99.91%
Solid-Liquid Mixture
890405-51-3
Research
25 mg
O=C1OC(COC2=C([N+]([O-])=O)C=CC=C2)CC1CC3=CC=CC=C3
327.33
Research
Cancer-Kinase/protease
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3BDO, MedChemExpress