missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 3D Tank Liner, Aegis™ 5-14 film, top and bottom drain

Product Code. 17840597 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
Unit Size:
Pack of 5
Pack of 8
This item is not returnable. View return policy

Product Code. 17840597

Brand: Thermo Scientific™ SH30988.01

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific 3D Tank Liners are composed of Aegis 5-14 film with dimensions optimized for Thermo Scientific drums and commonly-used, industry-standard cylindrical tanks. The tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.

  • Minimizes bioburden due to construction under sterile cGMP conditions
  • Helps eliminate post-use cleaning steps required with reusable containers, reducing cross-contamination risks and reduce cycle times
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 50 L
Quantity 50 L
Tubing Style C-Flex
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.