missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 3D Tank Liner, Aegis™ 5-14 film, top and bottom drain
Shop All Thermo Scientific Products

Click to view available options
Capacity (Metric):
50 L
100 L
200 L
Unit Size:
Pack of 5
Pack of 8
Description
- Minimizes bioburden due to construction under sterile cGMP conditions
- Helps eliminate post-use cleaning steps required with reusable containers, reducing cross-contamination risks and reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
Specifications
| Capacity (Metric) | 100 L |
| Quantity | 100 L |
| Tubing Style | C-Flex |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction