Learn More
Thermo Scientific™ 3K/4K Series Particle Counter Standards
Ensure accurate and convenient calibration of laser particle counters with Thermo Scientific™ 3K/4K Series Particle Counter Standards.
198.00 € - 420.00 €
Specifications
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
---|---|
Contents | Polymer particles in water |
Description | NIST traceable particles that provide calibration of laser particle counters |
Color | White or Colorless |
For Use With (Application) | Laser Particle Counter Calibration |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
11825012
|
Thermo Scientific™
3K1000 |
1.0 μm |
339.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11894992
|
Thermo Scientific™
3K-150 |
0.15 μm |
340.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11835012
|
Thermo Scientific™
3K1600 |
1.6 μm |
338.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11805002
|
Thermo Scientific™
3K-200 |
0.2 μm |
338.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11815002
|
Thermo Scientific™
3K-220 |
0.22 μm |
209.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11825002
|
Thermo Scientific™
3K-269 |
0.27 μm |
198.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11835002
|
Thermo Scientific™
3K-300 |
0.3 μm |
339.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11845002
|
Thermo Scientific™
3K-350 |
0.35 μm |
209.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11855002
|
Thermo Scientific™
3K-400 |
0.4 μm |
338.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11865002
|
Thermo Scientific™
3K-500 |
0.5 μm |
339.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11875002
|
Thermo Scientific™
3K-600 |
0.6 μm |
210.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11885002
|
Thermo Scientific™
3K-700 |
0.7 μm |
339.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11895002
|
Thermo Scientific™
3K-800 |
0.8 μm |
338.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11805012
|
Thermo Scientific™
3K-900 |
0.9 μm |
210.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11815012
|
Thermo Scientific™
3K-990 |
0.99 μm |
339.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11899722
|
Thermo Scientific™
4K-02 |
2.0 μm |
418.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11583253
|
Thermo Scientific™
4K-02 |
2.0 μm | N/A | N/A | N/A | |||||
11593253
|
Thermo Scientific™
4K-03 |
3 μm | N/A | N/A | N/A | |||||
11809732
|
Thermo Scientific™
4K-03 |
3 μm |
417.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11819732
|
Thermo Scientific™
4K-04 |
4 μm |
234.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11503263
|
Thermo Scientific™
4K-04 |
4 μm | N/A | N/A | N/A | |||||
11829732
|
Thermo Scientific™
4K-05 |
5.0 μm |
416.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11839732
|
Thermo Scientific™
4K-06 |
6 μm |
223.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11849732
|
Thermo Scientific™
4K-07 |
7 μm |
416.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11543263
|
Thermo Scientific™
4K-10 |
10 μm |
419.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11543273
|
Thermo Scientific™
4K100 |
100 μm | N/A | N/A | N/A | |||||
11859742
|
Thermo Scientific™
4K100 |
100 μm |
420.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11869732
|
Thermo Scientific™
4K-15 |
15 μm |
419.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11879732
|
Thermo Scientific™
4K-20 |
20 μm |
418.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11563263
|
Thermo Scientific™
4K-20 |
20 μm | N/A | N/A | N/A | |||||
11889732
|
Thermo Scientific™
4K-25 |
25 μm |
418.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11573263
|
Thermo Scientific™
4K-25 |
25 μm | N/A | N/A | N/A | |||||
11899732
|
Thermo Scientific™
4K-30 |
30 μm |
234.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11583263
|
Thermo Scientific™
4K-30 |
30 μm | N/A | N/A | N/A | |||||
11809742
|
Thermo Scientific™
4K-40 |
40 μm |
419.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11593263
|
Thermo Scientific™
4K-40 |
40 μm | N/A | N/A | N/A | |||||
11819742
|
Thermo Scientific™
4K-50 |
50 μm |
419.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11503273
|
Thermo Scientific™
4K-50 |
50 μm | N/A | N/A | N/A | |||||
11829742
|
Thermo Scientific™
4K-60 |
60 μm |
416.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11513273
|
Thermo Scientific™
4K-60 |
60 μm | N/A | N/A | N/A | |||||
11839742
|
Thermo Scientific™
4K-70 |
70 μm |
419.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11523273
|
Thermo Scientific™
4K-70 |
70 μm | N/A | N/A | N/A | |||||
11849742
|
Thermo Scientific™
4K-80 |
80 μm |
417.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11533273
|
Thermo Scientific™
4K-80 |
80 μm | N/A | N/A | N/A | |||||
11884992
|
Thermo Scientific™
3K-100 |
0.1 μm |
339.00 €
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Descripción
Ensure accurate and convenient calibration of laser particle counters with Thermo Scientific™ 3K/4K Series Particle Counter Standards. Compared with our 3000 and 4000 Series products, this product line is precisely diluted for immediate use in laser particle counters with minimal time-consuming concentration adjustments.
Features:- Suspensions of monodisperse polystyrene spheres for use in airborne or liquid particle dispersion systems
- Particle diameters traceable to the Standard Meter through the National Institute of Standards and Technology (NIST)
- Accurately calibrates or checks performance of laser particle counters used in cleanrooms and other contamination monitoring applications
- Particles prepared as low residue aqueous suspensions for minimal background interference
Especificaciones
Contains trace amount of surfactant to inhibit agglomeration and promote stability | |
NIST traceable particles that provide calibration of laser particle counters | |
Laser Particle Counter Calibration | |
Count Control Particles | |
15 mL |
Polymer particles in water | |
White or Colorless | |
Polystyrene | |
Certificate of Calibration and Traceability to NIST | |
1.59 at 589nm (25°C) |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.